Learn More
BINDER™ Temperature Analog Output
SDP
Catalog No. 08111574 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
BINDER Chambers BD 23, BD 400
Catalog No. For Use With (Equipment)
08-111-574 BINDER Chambers BD 23, BD 400
1 options
Catalog No. 08-111-574 Supplier BINDER™ Supplier No. 80120432
Add to Cart
Add to Cart
For Use With: BINDER units BD 23, BD 400
  • Output not adjustable
  • 4 to 20 mA
Specifications
Description Analog output 4-20 mA - for temperature values (output not adjustable)
For Use With (Equipment) BINDER Chambers BD 23, BD 400
Material Plastic
Type Analog Output
For Use With (Application) Temperature