Learn More
Metrohm USA TEMPERATURE BLK CABLE ADAPTER
SDP
Catalog No. NC0234703 Shop All Metrohm USA Products
Encompass
For connecting plug B (4 mm) to 2 mm socket.
Catalog No. NC0234703 Supplier Metrohm USA Supplier No. 62103140
For connecting plug B (4 mm) to 2 mm socket.