Learn More
Thermo Scientific™ Temperature Calibration for Cold Storage Devices, ISO 17025 PROMO
Shop All Thermo Scientific ProductsDescription
Temperature calibration for cold storage devices
- ISO 17025: Accredited certificate with data and uncertainties
- Regulatory requirements: Ideal for organizations in highly regulated environments where uncertainties are required to assure compliance.
- Included in service delivery: Found as left calibration data; TUR calculation Greater than 4:1 uncertainty ratio; Detailed calibration label; If a unit is out of tolerance an adjustment is performed
- Included on accredited certificate: Serial/asset number; Measurement data; Environmental conditions; Reported uncertainties (K=2); Conformity assessment; Reference equipment used
Specifications
Specifications
| For Use With (Equipment) | Cold Storage Devices |
| Duration | 1 Day |
| Product Type | Calibration |