Learn More
Thermo Scientific™ Temperature Calibration for Cold Storage Devices, ISO 17025
Catalog No. CALPRCOLD Shop All Thermo Scientific Products
Change view
Click to view available options
Duration:
1 Day
Catalog No. Duration
CALPRCOLD 1 Day
1 options
Catalog No. CALPRCOLD Supplier Thermo Scientific™ Supplier No. CALPRCOLD
Add to Cart
Add to Cart
Temperature calibration for cold storage devices. ISO 17025: Certificate with before and after data and uncertainties. Regulatory requirements: Ideal for highly regulated environments where uncertainties are required to assure compliance.
Temperature calibration for cold storage devices
  • ISO 17025: Accredited certificate with data and uncertainties
  • Regulatory requirements: Ideal for organizations in highly regulated environments where uncertainties are required to assure compliance.
  • Included in service delivery: Found as left calibration data; TUR calculation Greater than 4:1 uncertainty ratio; Detailed calibration label; If a unit is out of tolerance an adjustment is performed
  • Included on accredited certificate: Serial/asset number; Measurement data; Environmental conditions; Reported uncertainties (K=2); Conformity assessment; Reference equipment used
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Cold Storage Devices
Duration 1 Day
Product Type Calibration