Learn More
Perkin Elmer US LLC Temperature Calibration Kit
Supplier:  Perkin Elmer US LLC N5330318
Encompass_Preferred
The Temperature Calibration Kit is used to calibrate furnace temperatures in the DMA 8000.
Catalog No. 50-211-3462