Learn More
Saf T Pak Inc Sepio 9-2.52L Refrigerated (2-8C) Insulated Shipping Solution
SDP
Catalog No. 501904596 Shop All Saf-T-Pak Inc. Products
Encompass_Preferred
The cost-effective SEPIO is a tested system offering a low barrier of entry when shipping payloads needing refrigeration, & can be used as an Overpack when transporting Category A Infectious Substances (UN 2814 or UN 2900).FEATURES: Key Features | ClaimsPre-qualified for 2C to 8C & 15C to 25C using the same brick cassettesInmark Legacy ProfilesSeasonal and; Universal pack-outsCorrugated Cassettes for Easy HandlingMultiple payload sizesCost-effective solutionsWater-based refrigerant cassettesGlobal availability Temp/Profile/Duration:2-8/US48/UW72Summer (2-8C)Outside Dimensions - Shipping Carton(mm) 352 x 352 x 374(in) 13.9 x 13.9 x 14.7Inside Dimensions - Product Space(mm) 191 x 178 x 51(in) 7.5 x 7.0 x 2.0Dead Weight (Inc. Refrigerant)(kg) 4.9(lb) 10.9 Winter (2-8C) Outside Dimensions - Shipping Carton(mm) 352 x 352 x 374(in) 13.9 x 13.9 x 14.7 Inside Dimensions - Prod. Space(mm) 191 x 178 x 51(in) 7.5 x 7.0 x 2.0
Catalog No. 50-190-4596 Supplier Saf T Pak Inc Supplier No. 24391
The cost-effective SEPIO is a tested system offering a low barrier of entry when shipping payloads needing refrigeration, & can be used as an Overpack when transporting Category A Infectious Substances (UN 2814 or UN 2900).FEATURES: Key Features | ClaimsPre-qualified for 2C to 8C & 15C to 25C using the same brick cassettesInmark Legacy ProfilesSeasonal and; Universal pack-outsCorrugated Cassettes for Easy HandlingMultiple payload sizesCost-effective solutionsWater-based refrigerant cassettesGlobal availability Temp/Profile/Duration:2-8/US48/UW72Summer (2-8C)Outside Dimensions - Shipping Carton(mm) 352 x 352 x 374(in) 13.9 x 13.9 x 14.7Inside Dimensions - Product Space(mm) 191 x 178 x 51(in) 7.5 x 7.0 x 2.0Dead Weight (Inc. Refrigerant)(kg) 4.9(lb) 10.9 Winter (2-8C) Outside Dimensions - Shipping Carton(mm) 352 x 352 x 374(in) 13.9 x 13.9 x 14.7 Inside Dimensions - Prod. Space(mm) 191 x 178 x 51(in) 7.5 x 7.0 x 2.0