Learn More
Waters Corp Waters Temperature Control Module, Microprocessor-Controlled, Simultaneous 3-Module Control, Dual Communication
Catalog No. NC2868617 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Temperature Control Module II is a microprocessor-controlled unit for HPLC systems. It independently regulates and maintains the temperature of up to three column heater modules, ensuring stable operating conditions and insulating them from ambient changes.
  • Independent temperature setting for each connected column heater.
  • Alarms and on-screen error messages for temperature excursions.
  • Local mode keypad control or remote mode via system software.
  • Microprocessor-controlled for precise regulation and rapid stabilization.
  • Simultaneously controls up to three column heater modules.
  • Dual communication interfaces (Ethernet, IEEE-488).
  • Safety alarms and automatic shutdown for out-of-range temperatures.
  • Operates from 4°C to 40°C with 10% to 90% noncondensing humidity.
Catalog No. NC2868617 Supplier Waters Corp Supplier No. 186002441
The Temperature Control Module II is a microprocessor-controlled unit for HPLC systems. It independently regulates and maintains the temperature of up to three column heater modules, ensuring stable operating conditions and insulating them from ambient changes.
  • Independent temperature setting for each connected column heater.
  • Alarms and on-screen error messages for temperature excursions.
  • Local mode keypad control or remote mode via system software.
  • Microprocessor-controlled for precise regulation and rapid stabilization.
  • Simultaneously controls up to three column heater modules.
  • Dual communication interfaces (Ethernet, IEEE-488).
  • Safety alarms and automatic shutdown for out-of-range temperatures.
  • Operates from 4°C to 40°C with 10% to 90% noncondensing humidity.