Learn More
Braintree Scientific Temperature Controller 3 ft. cable only
SDP
Catalog No. 501956031
Encompass
has to be calibrated with the TCAT-2LV OR TCAT-2DF CANNOT BE USED WITH TCAT-2AC Only has a plug to the controller. Does not plug into wall as well. **Sold for research purposes only and not for human use**
Catalog No. 50-195-6031 Supplier Braintree Scientific Supplier No. TCATCB3A
has to be calibrated with the TCAT-2LV OR TCAT-2DF CANNOT BE USED WITH TCAT-2AC Only has a plug to the controller. Does not plug into wall as well. **Sold for research purposes only and not for human use**