Learn More
Chemglass Life Sciences Dual Temperature Controller, J-Kem, Apollo, Type "K" (-50°C to 1200°C), Complete
SDP
Catalog No. 50401835
Encompass_Preferred
Dual Temperature Controller, J-Kem, Apollo, Type "K" (-50C to 1200C), Complete. J-Kem Scientific Apollo dual temperature controller features two independent temperature controllers in a single cabinet. Each independent channel has its own high power outlet, 100 hour timer to turn heating/cooling on or off at a user set time, and solid state over temperature protection circuit. Temperature ramping and serial communications are also provided with this unit.PLEASE NOTE: THE TYPE T, J AND K CONTROLLERS ARE SUPPLIED COMPLETE WITH: 1/8 DIAMETER X 12 LONG PTFE COATED THERMOCOUPLE, 10FT COILED EXTENSION CORD, AND A 14/20 PTFE THERMOCOUPLE ADAPTER. THE RTD UNIT IS THE CONTROLLER ONLY.Specifications:Dimensions: 4" H x 8" W x 9" DPower: 10 Amps @ 120VAC, 1200 Watts MAX/Channel, 1800 Watts (Total)
Catalog No. 50-401-835 Supplier Chemglass Life Sciences Supplier No. CG321003
Dual Temperature Controller, J-Kem, Apollo, Type "K" (-50C to 1200C), Complete. J-Kem Scientific Apollo dual temperature controller features two independent temperature controllers in a single cabinet. Each independent channel has its own high power outlet, 100 hour timer to turn heating/cooling on or off at a user set time, and solid state over temperature protection circuit. Temperature ramping and serial communications are also provided with this unit.PLEASE NOTE: THE TYPE T, J AND K CONTROLLERS ARE SUPPLIED COMPLETE WITH: 1/8 DIAMETER X 12 LONG PTFE COATED THERMOCOUPLE, 10FT COILED EXTENSION CORD, AND A 14/20 PTFE THERMOCOUPLE ADAPTER. THE RTD UNIT IS THE CONTROLLER ONLY.Specifications:Dimensions: 4" H x 8" W x 9" DPower: 10 Amps @ 120VAC, 1200 Watts MAX/Channel, 1800 Watts (Total)