Learn More
Chemglass Life Sciences Temperature Controller, J-Kem, Model 270, Type "J" (0°C to 800°C), Complete
SDP
Supplier:  Chemglass Life Sciences CG320702
Encompass_Preferred
Temperature Controller, J-Kem, Model 270, Type "J" (0C to 800C), Complete. J-Kem Scientific microprocessor temperature controller. Comes with a completely INDEPENDENT BACK UP controller to guard against accidents from equipment failure.The desired temperature is entered into the digital display of the setpoint controller which regulates heating. A high temperature limit is then entered into the second digital back-up. If the reaction reaches the setpoint of the second controller, power is shut off from the output circuit until the unit is manually reset. Both controllers independently monior the reaction temperature and in the event that one controller fails, the other takes over to prevent a accidentPLEASE NOTE: CONTROLLER IS SUPPLIED COMPLETE WITH: A DUAL ELEMENT 1/4 DIAMETER X 12 LONG PTFE COATED THERMOCOUPLE, A DUAL ELEMENT THERMOCOUPLE EXTENSION CORD AND A 14/20 PTFE THERMOCOUPLE ADAPTERSpecifications:Dimensions: 3.25" H x 7.5" W x 9" DPower: 15 Amps @ 120VAC, 1800 Watts
Catalog No. 50-401-828