Learn More
Chemglass Life Sciences Temperature Controller for CHEMcell Bioreactor Rocker Trays
SDP
Catalog No. 502733331
Encompass
J-KEM Temperature Controller used to control the temperature of the integral heating pad of the CHEMcell culture bag tray.The controller is used for both tray sizes, CLS-1200-200 and CLS-1200-300, as well as the 4 watt filter vent heater, CLS-1200-FH.
Catalog No. 50-273-3331 Supplier Chemglass Life Sciences Supplier No. CLS12002CH
J-KEM Temperature Controller used to control the temperature of the integral heating pad of the CHEMcell culture bag tray.The controller is used for both tray sizes, CLS-1200-200 and CLS-1200-300, as well as the 4 watt filter vent heater, CLS-1200-FH.