Learn More
Chemglass Life Sciences Temperature Controller, J-Kem, Model HCC with Clamp, Type "T" (-200°C to 250°C)
SDP
Supplier:  Chemglass Life Sciences CG197265
Encompass_Preferred
Temperature Controller, J-Kem, Model HCC with Clamp, Type "T" (-200C to 250C). The J-KEM Scientific HCC controller is designed to power large scale equipment such as 50L to 100L heating mantles. Designed with the same safety features as the Model 270, the HCC incorporates a 100 hour digital timer to turn heating on or off. PLEASE NOTE: THIS CONTROLLER REQUIRES ONE DUAL ELEMENT THERMOCOUPLE OR TWO SINGLE ELEMENT THERMOCOUPLES.
Catalog No. CG197265