Learn More
BINDER™ Temperature and Humidity Calibration Certificate
SDP
Catalog No. 502875768 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
KBF PRO Series
KBF Series
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
50-287-5768 KBF Series
50-287-5726 KBF PRO Series
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-287-5768 Supplier BINDER™ Supplier No. 80122577
Add to Cart
Add to Cart
Calibration certificate including 27 temperature measurements and 1 humidity measurement.
  • 27 temperature measuring points
  • 1 humidity measuring point, at 25°C/60% RH, or at specified values
Specifications
For Use With (Equipment) KBF Series
Certifications/Compliance DIN12880
Duration 1 Day
Product Type Calibration Certificate