Learn More
Research Products International Corp Temperature and Humidity Monitor, Supplied with Battery, 4 1/4 x 3 3/4 x 7/8 Inches
SDP
Catalog No. 50550350
Encompass
View both temperature and humidity readings even from across a room with this extra large display. Convenient for use in labs, storage areas, incubators, refrigerators or fume hoods. Features automatic recall of min/max readings and manual recall.
Supplied with AAA battery that will last up to one year.
Catalog No. 50-550-350 Supplier Research Products International Corp Supplier No. 800016
View both temperature and humidity readings even from across a room with this extra large display. Convenient for use in labs, storage areas, incubators, refrigerators or fume hoods. Features automatic recall of min/max readings and manual recall.
Supplied with AAA battery that will last up to one year.