Learn More
Grainger Extech Temperature PID Controller, 1/16 DIN, 4-20 mA Output, 1 ea
Catalog No. 502756729
Encompass_Preferred
This 1/16 DIN temperature PID controller utilizes Fuzzy Logic enhanced PID control to eliminate overshoot, unwanted process fluctuations, and drift. The unit features a Soft-Start function designed to protect heaters during cold starts by providing exact, slow heating, making it ideal for processes such as those in the thermo-plastics industry. It is compatible with various thermocouple and RTD inputs and supports a single-stage Ramp and Soak program with a Ramp-to-Setpoint limit.
  • Fuzzy Logic enhanced PID control
  • Soft-Start feature for heater protection
  • Accepts thermocouple and RTD inputs
  • Dual 4-digit LED displays for process and setpoint values
  • Single stage Ramp and Soak program
  • Two latching alarm relays with 8 alarm modes
  • One-touch auto tuning for quick setup
  • Manual override mode
  • Includes mounting hardware and screw terminals
Catalog No. 50-275-6729 Supplier Grainger Supplier No. 15E601
This 1/16 DIN temperature PID controller utilizes Fuzzy Logic enhanced PID control to eliminate overshoot, unwanted process fluctuations, and drift. The unit features a Soft-Start function designed to protect heaters during cold starts by providing exact, slow heating, making it ideal for processes such as those in the thermo-plastics industry. It is compatible with various thermocouple and RTD inputs and supports a single-stage Ramp and Soak program with a Ramp-to-Setpoint limit.
  • Fuzzy Logic enhanced PID control
  • Soft-Start feature for heater protection
  • Accepts thermocouple and RTD inputs
  • Dual 4-digit LED displays for process and setpoint values
  • Single stage Ramp and Soak program
  • Two latching alarm relays with 8 alarm modes
  • One-touch auto tuning for quick setup
  • Manual override mode
  • Includes mounting hardware and screw terminals