Learn More
Corning™ Temperature Probe Support Clamp for Hot Plate Stirrers Produced in China
Catalog No. 08757300 Shop All Corning Products
Change view
Click to view available options
For Use With (Equipment):
Corning 6795PR and 400188
Catalog No. For Use With (Equipment)
08-757-300 Corning 6795PR and 400188
1 options
Catalog No. 08-757-300 Supplier Corning™ Supplier No. 440141CH
Add to Cart
Add to Cart
Support clamp for temperature probes designed to be used with boss head clamp.
Corning offers a 7/16 in. (16.5 cm) support clamp for Corning Cat. No. 6795PR and 400188 temperature probes.
  • Support has two openings, 1/3 in. (9 mm) and 0.15 in. (3.75 mm), for holding probes.
  • Designed to be used with a boss head clamp (Corning Cat. No. 440140) or standard right angle clamp.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Rod 7/16 in.
Product Type Support Clamp
For Use With (Equipment) Corning 6795PR and 400188
Quantity 1 Ea.