Learn More
World Precision Instrument Temperature Sensor
SDP
Catalog No. 502538742
Encompass
This temperature sensor is based on a 2.0 mm tip diameter high quality miniature platinum RTD (Resistance Temperature Detector) electrode. This design has been shown to provide greater accuracy, stability and interchangeability during temperature measurements than traditional thermistor and thermocouple sensors.
Catalog No. 50-253-8742 Supplier World Precision Instrument Supplier No. ISOTEMP2
This temperature sensor is based on a 2.0 mm tip diameter high quality miniature platinum RTD (Resistance Temperature Detector) electrode. This design has been shown to provide greater accuracy, stability and interchangeability during temperature measurements than traditional thermistor and thermocouple sensors.