Learn More
Analytical Sales & Services Temperature Transfer Cover for 24-Well, 18mm Spacing Reactors for 15mm OD Vials
Catalog No. 5000804492
Encompass
Temperature Transfer Cover for 24-Well, 18mm Spacing Reactors for 15mm OD Vials\n\nWhen using 15mm OD, 1 Dram vials in 24-well reactor blocks, this cover is perfect for those who prefer to cap their vials instead of sealing them with the standard aluminum cover, rubber mat and PFA film that come with the block assembly. When used in conjunction with a Thermal Transfer Deck (TTD) and external liquid chiller, the cover helps control excessive heat generated by a Lumidox II LED array. Its 17mm diameter, 20.5mm deep wells ensure the highest aluminum-to-aluminum ratio for maximum heat dispersal.\n\nThe vials in the reactor block provide the alignment for the cover, but for extra stability users can secure the cover with twelve (12) screws which are in the same arrangement as a standard flat lid. The cover can also be flipped over and used as a reactor in itself or vial tray.\n\n 
Catalog No. 50-008-04492 Supplier Analytical Sales & Services Supplier No. 24122
Temperature Transfer Cover for 24-Well, 18mm Spacing Reactors for 15mm OD Vials\n\nWhen using 15mm OD, 1 Dram vials in 24-well reactor blocks, this cover is perfect for those who prefer to cap their vials instead of sealing them with the standard aluminum cover, rubber mat and PFA film that come with the block assembly. When used in conjunction with a Thermal Transfer Deck (TTD) and external liquid chiller, the cover helps control excessive heat generated by a Lumidox II LED array. Its 17mm diameter, 20.5mm deep wells ensure the highest aluminum-to-aluminum ratio for maximum heat dispersal.\n\nThe vials in the reactor block provide the alignment for the cover, but for extra stability users can secure the cover with twelve (12) screws which are in the same arrangement as a standard flat lid. The cover can also be flipped over and used as a reactor in itself or vial tray.\n\n 