Learn More
Chemglass Life Sciences Temperature Controller, J-Kem, Type "T", -200 to 250°C, Model 260
SDP
Catalog No. 501531618
Encompass_Preferred
Temperature Controller, J-Kem, Type "T", -200 to 250C, Model 260. The J-Kem Scientific microprocessor based temperature controller is for use with the CG-1966 and CG-1967 pilot plant reactors. The Model 260 disconnects all output power if the process temperature exceeds the entered temperature by a user-specified amount, or following recovery from a power failure. The outputs remain disconnected until manually reset. Over temperature condition is signaled by an indicator lamp and audible alarm. Controller has two outlets on the front for connection to heaters. The Model 260 controls temperature to within 0.1C of liquid. NOTE: Support rod, thermocouple and extension cord are sold separately. Specifications: Dimensions H x W x D (in): 3.25 x 7.5 x 9. Power: 15 amps @ 120 VAC, 1800 Watts.
Catalog No. 50-153-1618 Supplier Chemglass Life Sciences Supplier No. CG320591
Temperature Controller, J-Kem, Type "T", -200 to 250C, Model 260. The J-Kem Scientific microprocessor based temperature controller is for use with the CG-1966 and CG-1967 pilot plant reactors. The Model 260 disconnects all output power if the process temperature exceeds the entered temperature by a user-specified amount, or following recovery from a power failure. The outputs remain disconnected until manually reset. Over temperature condition is signaled by an indicator lamp and audible alarm. Controller has two outlets on the front for connection to heaters. The Model 260 controls temperature to within 0.1C of liquid. NOTE: Support rod, thermocouple and extension cord are sold separately. Specifications: Dimensions H x W x D (in): 3.25 x 7.5 x 9. Power: 15 amps @ 120 VAC, 1800 Watts.