Learn More
Electron Microscopy Sciences Tempfix Adhesive Set
SDP
Catalog No. 50980327
Encompass_Preferred
A thermoplastic adhesive for mounting powder specimens and small particles for SEM. It does not contain any solvents and it is stable in high vacuum. It is not sticky at room temperature but becomes adhesive at 40 degree C and melts at 120 degree C. Tempfix may also be used as an embedding medium. EMS introduces a new reliable line of conductive adhesives which are certified by the Bureau Veritas Quality International. Quality Standards: ISO 9001; EN29001; ANSI/ASQC Q91
Catalog No. 50-980-327 Supplier Electron Microscopy Sciences Supplier No. 12668
A thermoplastic adhesive for mounting powder specimens and small particles for SEM. It does not contain any solvents and it is stable in high vacuum. It is not sticky at room temperature but becomes adhesive at 40 degree C and melts at 120 degree C. Tempfix may also be used as an embedding medium. EMS introduces a new reliable line of conductive adhesives which are certified by the Bureau Veritas Quality International. Quality Standards: ISO 9001; EN29001; ANSI/ASQC Q91