Learn More
USA Scientific Inc TempPlate pierceable sealing foil, sterile
Catalog No. NC9972543
Encompass
TempPlate pierceable aluminum sealing foil, -80C to +120C, sterile, 50/box
Catalog No. NC9972543 Supplier USA Scientific Inc Supplier No. 29230110
TempPlate pierceable aluminum sealing foil, -80C to +120C, sterile, 50/box