Learn More
SKC Inc Wipe Template, Paper, 10x10 cm, 250/pack
SDP
Catalog No. NC9536923
Encompass
SKC Wipe Template for surface sampling, disposable manila paper, 10 X 10 cm, (pkg/250) DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”
Catalog No. NC9536923 Supplier SKC Inc Supplier No. 2252415
SKC Wipe Template for surface sampling, disposable manila paper, 10 X 10 cm, (pkg/250) DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”