Learn More
American Biotech Supply TEMPLOG LAB FREEZER 30CF
Catalog No. NC2389381
Encompass_Preferred
TempLog Premier Manual Defrost Laboratory Freezers are purpose built for scientific and medical applications. Units feature a digital microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies. A manual defrost design prevents product damaging temperature spikes from defrost cycles.
Catalog No. NC2389381 Supplier American Biotech Supply Supplier No. ABTMFP30TS
Add to Cart
Add to Cart
Request a Quote
TempLog Premier Manual Defrost Laboratory Freezers are purpose built for scientific and medical applications. Units feature a digital microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies. A manual defrost design prevents product damaging temperature spikes from defrost cycles.