Learn More
Medchemexpress LLC Tempo | 2564-83-2 | 156.25 | 25 G
SDP
Catalog No. 5000450019
Encompass_Preferred
Tempo is a nitric oxide radical and a selective scavenger of reactive oxygen species (ROS) in mitochondria. It also functions as an organocatalyst that disproportionates superoxide and oxidizes primary alcohols to aldehydes in a catalytic cycle. Tempo has mutagenic and antioxidant effects, can induce DNA strand breaks, and exhibits cytotoxic and mutagenic properties in mouse lymphoma cells.
  • Selective scavenger of reactive oxygen species (ROS)
  • Organocatalyst for superoxide disproportionation
  • Oxidizes primary alcohols to aldehydes
  • Exhibits mutagenic and antioxidant effects
  • Induces DNA strand breaks
  • Cytotoxic and mutagenic in mouse lymphoma cells
Catalog No. 50-004-50019 Supplier Medchemexpress LLC Supplier No. 25GHYW001187
Tempo is a nitric oxide radical and a selective scavenger of reactive oxygen species (ROS) in mitochondria. It also functions as an organocatalyst that disproportionates superoxide and oxidizes primary alcohols to aldehydes in a catalytic cycle. Tempo has mutagenic and antioxidant effects, can induce DNA strand breaks, and exhibits cytotoxic and mutagenic properties in mouse lymphoma cells.
  • Selective scavenger of reactive oxygen species (ROS)
  • Organocatalyst for superoxide disproportionation
  • Oxidizes primary alcohols to aldehydes
  • Exhibits mutagenic and antioxidant effects
  • Induces DNA strand breaks
  • Cytotoxic and mutagenic in mouse lymphoma cells