Learn More
Apexbio Technology LLC Tempo 2564-83-2 200mg
SDP
Catalog No. 5000089815
Encompass_Preferred
Tempo (CAS 2564-83-2) is a stable nitroxide radical widely employed in biomedical research for its redox-active properties It facilitates the dismutation of superoxide radicals thereby modulating mitochondrial reactive oxygen species (ROS) levels and influencing cellular oxidative stress responses Tempo also serves as an oxidation catalyst particularly in the transformation of primary alcohols to aldehydes Studies indicate that Tempo exerts both antioxidant and mutagenic effects inducing DNA strand breaks and displaying cytotoxic and mutagenic activities in in vitro models such as murine lymphoma cells It is utilized for investigating oxidative stress redox regulation and chemical mutagenesis
Catalog No. 50-000-89815 Supplier Apexbio Technology LLC Supplier No. C6812200
Tempo (CAS 2564-83-2) is a stable nitroxide radical widely employed in biomedical research for its redox-active properties It facilitates the dismutation of superoxide radicals thereby modulating mitochondrial reactive oxygen species (ROS) levels and influencing cellular oxidative stress responses Tempo also serves as an oxidation catalyst particularly in the transformation of primary alcohols to aldehydes Studies indicate that Tempo exerts both antioxidant and mutagenic effects inducing DNA strand breaks and displaying cytotoxic and mutagenic activities in in vitro models such as murine lymphoma cells It is utilized for investigating oxidative stress redox regulation and chemical mutagenesis