Learn More
Medchemexpress LLC Tempo | 2564-83-2 | 99.86% | 156.25 | 100 MG
SDP
Catalog No. 5000364818
Encompass_Preferred
Tempo is a nitric oxide radical and a selective scavenger of reactive oxygen species (ROS) in mitochondria. It also functions as an organocatalyst that disproportionates superoxide and oxidizes primary alcohols to aldehydes in a catalytic cycle. Tempo has mutagenic and antioxidant effects, and it can induce DNA strand breaks. It also exerts cytotoxic and mutagenic properties in mouse lymphoma cells.
  • Nitric oxide radical and selective scavenger of ROS in mitochondria
  • Functions as an organocatalyst that disproportionates superoxide
  • Oxidizes primary alcohols to aldehydes in a catalytic cycle
  • Has mutagenic and antioxidant effects
  • Can induce DNA strand breaks
  • Exerts cytotoxic and mutagenic properties in mouse lymphoma cells
Catalog No. 50-003-64818 Supplier Medchemexpress LLC Supplier No. 100MGHYW001187
Add to Cart
Add to Cart
Tempo is a nitric oxide radical and a selective scavenger of reactive oxygen species (ROS) in mitochondria. It also functions as an organocatalyst that disproportionates superoxide and oxidizes primary alcohols to aldehydes in a catalytic cycle. Tempo has mutagenic and antioxidant effects, and it can induce DNA strand breaks. It also exerts cytotoxic and mutagenic properties in mouse lymphoma cells.
  • Nitric oxide radical and selective scavenger of ROS in mitochondria
  • Functions as an organocatalyst that disproportionates superoxide
  • Oxidizes primary alcohols to aldehydes in a catalytic cycle
  • Has mutagenic and antioxidant effects
  • Can induce DNA strand breaks
  • Exerts cytotoxic and mutagenic properties in mouse lymphoma cells