Learn More
Medchemexpress LLC HY-100561 200mg Medchemexpress, Tempol CAS:2226-96-2 Purity:>98%
SDP
Catalog No. 501967748
Encompass_Preferred
Medchemexpress, HY-100561 200mg Tempol CAS:2226-96-2 Tempol is a general superoxide dismutase (SOD)-mimetic drug that efficiently neutralizes reactive oxygen species (ROS). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7748 Supplier Medchemexpress LLC Supplier No. HY100561200MG
Medchemexpress, HY-100561 200mg Tempol CAS:2226-96-2 Tempol is a general superoxide dismutase (SOD)-mimetic drug that efficiently neutralizes reactive oxygen species (ROS). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.