Learn More
Medchemexpress LLC Tempol-d17,15N | 90429-66-6 | 98.4% | 190.34 | C9HD1715NO2 | 10 MG
SDP
Catalog No. 5000212689
Encompass_Preferred
Tempol-d17,15N is the deuterium- and 15N-labeled analogue of Tempol (4-hydroxy-TEMPO), intended for research use as a stable isotope tracer and quantitative internal standard. The parent nitroxide functions as a superoxide dismutase (SOD)-mimetic that neutralizes reactive oxygen species, and isotopic labeling enables tracer studies and accurate quantitation in analytical workflows.
  • Deuterium and 15N labeled for use as a stable isotope tracer.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • High purity (98.4%) appropriate for analytical applications.
  • Acts as an SOD-mimetic nitroxide that neutralizes reactive oxygen species.
  • Available in small, weighed quantities for analytical and tracer studies.
Catalog No. 50-002-12689 Supplier Medchemexpress LLC Supplier No. HY100561S10MG
Tempol-d17,15N is the deuterium- and 15N-labeled analogue of Tempol (4-hydroxy-TEMPO), intended for research use as a stable isotope tracer and quantitative internal standard. The parent nitroxide functions as a superoxide dismutase (SOD)-mimetic that neutralizes reactive oxygen species, and isotopic labeling enables tracer studies and accurate quantitation in analytical workflows.
  • Deuterium and 15N labeled for use as a stable isotope tracer.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • High purity (98.4%) appropriate for analytical applications.
  • Acts as an SOD-mimetic nitroxide that neutralizes reactive oxygen species.
  • Available in small, weighed quantities for analytical and tracer studies.