Learn More
Enzo Life Sciences TEMPOL, (1g), CAS Number: 2226-96-2
SDP
Supplier:  Enzo Life Sciences ALX430081G001
Encompass_Preferred
SOD mimetic. Small molecule cell-permeable superoxide dismutase (SOD) mimetic. It attenuates superoxide anion and peroxynitrite-induced inflammation, lowers blood peressure in a variety of models and displays neuroprotective effects. It restores mitochondrial and cardiac functions in TNFα-induced oxidative stress and reduces cardiac hypertrophy in chronic hypoxic rats. Alternative Name: 4-OH-TEMPO. 4-Hydroxy-TEMPO. 4-Hydroxy-2,2,6,6-tetramethylpiperidinyloxy, free radical. Formula: C9H18NO2. MW: 172.2. CAS Number: 2226-96-2. Purity: ≥98% (HPLC). Appearance: Yellow to orange crystalline solid. Solubility: Soluble in water, 100% ethanol, methanol or DMSO. Long Term Storage: -20°C
Catalog No. NC0748955