Learn More
Medchemexpress LLC Tempone-15N,d16 (4-oxo-Tempo-15N,d16) | 80404-14-4 | 1 MG
SDP
Catalog No. 5000339725
Encompass_Preferred
Tempone-15N,d16 is the deuterium and 15N labeled Tempone, serving as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during drug development. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer and internal standard
  • Suitable for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium and 15N labeled compound
  • Solid appearance, light yellow to yellow color
  • Soluble in DMSO (requires ultrasonic and warming)
Catalog No. 50-003-39725 Supplier Medchemexpress LLC Supplier No. HY129242S1MG
Tempone-15N,d16 is the deuterium and 15N labeled Tempone, serving as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during drug development. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer and internal standard
  • Suitable for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium and 15N labeled compound
  • Solid appearance, light yellow to yellow color
  • Soluble in DMSO (requires ultrasonic and warming)