Learn More
Medchemexpress LLC Tempone (4-Oxo-Tempo) | 2896-70-0 | 99.1% | 25 G
SDP
Catalog No. 5000342811
Encompass_Preferred
Tempone (4-Oxo-Tempo) is a stable water-soluble nitro radical used as a contrast agent for metabolic activity and hypoxic sensitivity in electron spin resonance spectroscopy, magnetic resonance imaging, and dynamic nuclear polarization.
  • Reduces superoxide radicals by mimicking superoxide dismutase (SOD) activity
  • Decreases the formation of hydroxyl radicals and peroxynitrites
  • Applicable in studies of ischemia-reperfusion injury and acute renal failure
Catalog No. 50-003-42811 Supplier Medchemexpress LLC Supplier No. 25GHY129242
Tempone (4-Oxo-Tempo) is a stable water-soluble nitro radical used as a contrast agent for metabolic activity and hypoxic sensitivity in electron spin resonance spectroscopy, magnetic resonance imaging, and dynamic nuclear polarization.
  • Reduces superoxide radicals by mimicking superoxide dismutase (SOD) activity
  • Decreases the formation of hydroxyl radicals and peroxynitrites
  • Applicable in studies of ischemia-reperfusion injury and acute renal failure