Learn More
Enzo Life Sciences TEMPONE-H. hydrochloride (250mg). CAS: 3637-11-4
SDP
Supplier:  Enzo Life Sciences ALX430071M250
Encompass_Preferred
Very effective, cell permeable and non-toxic spin trap for the detection of superoxide radical and peroxynitrite. Alternative name: 1-Hydroxy-2,2,6,6-tetramethyl-4-oxo-piperidine . HCl. Purity: ≥99% (GLC). Solubility: Soluble in DMSO, water, methanol or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-9183