Learn More
Exergen Corporation Temporal Contact Thermometer ComfortScanner
SDP
Catalog No. NC1796602
Encompass_Preferred
The TemporalScanner Thermometer is a totally non-invasive system with advanced infrared technology providing maximum ease of use with quick, consistently accurate measurements. Advanced, patented technology measures temperatures with a gentle stroke across the forehead. It captures naturally emitted heat from the skin above the temporal artery, 1,000 times a second and then gives a reading of the most accurate temperature.
Catalog No. NC1796602 Supplier Exergen Corporation Supplier No. 140008
The TemporalScanner Thermometer is a totally non-invasive system with advanced infrared technology providing maximum ease of use with quick, consistently accurate measurements. Advanced, patented technology measures temperatures with a gentle stroke across the forehead. It captures naturally emitted heat from the skin above the temporal artery, 1,000 times a second and then gives a reading of the most accurate temperature.