Learn More
USA Scientific Inc TempPlate 384w PCR plate
Catalog No. 502745491
Encompass
TempPlate polypropylene 384-well PCR plate, white, barcoded on the A1-A24 side, A24 and P24 corner notches, 10/box.
Catalog No. 50-274-5491 Supplier USA Scientific Inc Supplier No. 14584690
TempPlate polypropylene 384-well PCR plate, white, barcoded on the A1-A24 side, A24 and P24 corner notches, 10/box.