Learn More
USA Scientific Inc TempPlate EXT sealing foil
Catalog No. 502744755
Encompass
TempPlate EXT sealing foil, standard cut for PCR and cold storage applications, non-str, 100/box
Catalog No. 50-274-4755 Supplier USA Scientific Inc Supplier No. 29980100
TempPlate EXT sealing foil, standard cut for PCR and cold storage applications, non-str, 100/box