Learn More
USA Scientific Inc TempPlate non-skirt PP 0.1 mL
Catalog No. 502745482
Encompass
TempPlate non-skirted polypropylene 0.1 mL 96-well PCR plate, low profile, H12 notch, white, 10/box
Catalog No. 50-274-5482 Supplier USA Scientific Inc Supplier No. 14028590
TempPlate non-skirted polypropylene 0.1 mL 96-well PCR plate, low profile, H12 notch, white, 10/box