Learn More
USA Scientific Inc TempPlate RT Select clear film
Catalog No. 502745107
Encompass
TempPlate RT Select optically clear real-time PCR film with strong silicone adhesive, -70C to +100C, str, 50/box
Catalog No. 50-274-5107 Supplier USA Scientific Inc Supplier No. 29217810
TempPlate RT Select optically clear real-time PCR film with strong silicone adhesive, -70C to +100C, str, 50/box