Learn More
USA Scientific Inc TempPlate sealing film,
Catalog No. 502745257
Encompass
TempPlate polypropylene sealing film, -40C to +120C, non-sterile, 100/box
Catalog No. 50-274-5257 Supplier USA Scientific Inc Supplier No. 29210000
TempPlate polypropylene sealing film, -40C to +120C, non-sterile, 100/box