Learn More
Andwin Scientific TEMPPLATE SEALING FOIL -70C
SDP
Supplier:  Andwin Scientific 29230110
Encompass_Preferred
The TempPlate pierceable sealing foil offers a durable seal without the use of heat sealing devices. The foil is pierceable and is free of RNase, DNase, and nucleic acid. This product can work within the temperature range of -80C to +120C. It measures 146.1 x 82.5 mm.
Catalog No. 50-820-088