Learn More
Andwin Scientific TEMPPLATE SEALING FOIL -70C
SDP
Catalog No. 50820087
Encompass_Preferred
TempPlate;Sealing Foil, -70C to +110C, pierce-able 100/PK
Catalog No. 50-820-087 Supplier Andwin Scientific Supplier No. 29230100
TempPlate;Sealing Foil, -70C to +110C, pierce-able 100/PK