Learn More
USA Scientific Inc TempPlate semi-skirt 96w PCR
Catalog No. 502745338
Encompass
TempPlate semi-skirted polypropylene 0.2 mL 96-well PCR plate, straight skirt, natural, barcoded on the H1-H12 side, A12 corner notch. 10/box.
Catalog No. 50-274-5338 Supplier USA Scientific Inc Supplier No. 14229200
TempPlate semi-skirted polypropylene 0.2 mL 96-well PCR plate, straight skirt, natural, barcoded on the H1-H12 side, A12 corner notch. 10/box.