Learn More
USA Scientific Inc TempPlate semiskirt PCR plate
Catalog No. 502744948
Encompass
TempPlate semi-skirted polypropylene 0.1 mL 96-well PCR plate, low profile, straight skirt, white, barcode on A1-A12 side, H12 corner notch. 10/box.
Catalog No. 50-274-4948 Supplier USA Scientific Inc Supplier No. 14229990
Add to Cart
Add to Cart
TempPlate semi-skirted polypropylene 0.1 mL 96-well PCR plate, low profile, straight skirt, white, barcode on A1-A12 side, H12 corner notch. 10/box.