Learn More
Perkin Elmer US LLC TempSET, without Probe
Catalog No. 502104239
Encompass_Preferred
The SPB TempSET is designed to shut-off SPB systems with no lab supervision. Ideal for overnight digestions and evaporations where samples require volume reductions (volume control 2.5 mL). Increase lab efficiency by automatically digesting samples off-hours.
Catalog No. 50-210-4239 Supplier Perkin Elmer US LLC Supplier No. N9300810
The SPB TempSET is designed to shut-off SPB systems with no lab supervision. Ideal for overnight digestions and evaporations where samples require volume reductions (volume control 2.5 mL). Increase lab efficiency by automatically digesting samples off-hours.