Learn More
Medchemexpress LLC HY-50910 10mg Medchemexpress, Temsirolimus CAS:162635-04-3 Purity:>98%
SDP
Catalog No. 502029355
Encompass_Preferred
Medchemexpress, HY-50910 10mg Temsirolimus CAS:162635-04-3 Temsirolimus is an inhibitor of mTOR with an IC50 of 1.76 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9355 Supplier Medchemexpress LLC Supplier No. HY5091010MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-50910 10mg Temsirolimus CAS:162635-04-3 Temsirolimus is an inhibitor of mTOR with an IC50 of 1.76 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.