Learn More
Apexbio Technology LLC Temsirolimus,10mg CAS# 162635-04-3
SDP
Catalog No. 502240061
Encompass_Preferred
CCI-779 is a potent inhibitor of mTOR with IC50 values of 0.6, 0.7, 0.7 and 50 nM for BT-474, MDA-MB-468, SKBR-3 and MCF-7 cells, respectively [1].CCI-779 is an ester derivative of rapamycin and has improved pharmaceutical properties. As a mTOR inhibitor, CCI-779 affected cell proliferation in cancer cells in which the cell cycle targets are dysregulated by mTOR. When treated with a panel of 8 human breast cancer cell lines, CCI-779 showed potent antigrowth activity with IC50 values of 0.6, 0.7 and 0.7nM for BT-474, MDA-MB-468 and SKBR-3 cells, respectively. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0061 Supplier Apexbio Technology LLC Supplier No. A831410
CCI-779 is a potent inhibitor of mTOR with IC50 values of 0.6, 0.7, 0.7 and 50 nM for BT-474, MDA-MB-468, SKBR-3 and MCF-7 cells, respectively [1].CCI-779 is an ester derivative of rapamycin and has improved pharmaceutical properties. As a mTOR inhibitor, CCI-779 affected cell proliferation in cancer cells in which the cell cycle targets are dysregulated by mTOR. When treated with a panel of 8 human breast cancer cell lines, CCI-779 showed potent antigrowth activity with IC50 values of 0.6, 0.7 and 0.7nM for BT-474, MDA-MB-468 and SKBR-3 cells, respectively. Other sizes are also available. Please inqury us for quote.