Learn More
Cayman Chemical Temsirolimus, Purity: ≥95%, CAS: 162635-04-3, 10 mg
SDP
Catalog No. 502952753
Encompass_Preferred
A dihydroxymethyl propionic acid ester of rapamycin with improved solubility that specifically inhibits mTOR signaling with a potency similar to that of rapamycin; demonstrates cytostatic activity in various mouse models of human tumors, including glioblastomas, prostate carcinoma, pancreatic, liver, breast cancers, and medulloblastoma
Catalog No. 50-295-2753 Supplier Cayman Chemical Supplier No. 1159010 MG
A dihydroxymethyl propionic acid ester of rapamycin with improved solubility that specifically inhibits mTOR signaling with a potency similar to that of rapamycin; demonstrates cytostatic activity in various mouse models of human tumors, including glioblastomas, prostate carcinoma, pancreatic, liver, breast cancers, and medulloblastoma