Learn More
Medchemexpress LLC Temsirolimus | 162635-04-3 | C56H87NO16 | 200MG
SDP
Catalog No. 5000358394
Encompass_Preferred
Temsirolimus is an inhibitor of mTOR with an IC50 of 1.76 μM. It activates autophagy and prevents deterioration of cardiac function in animal models. This product is an analytical standard intended for research and analytical applications.
  • Inhibits mTOR kinase activity
  • Activates autophagy
  • Prevents deterioration of cardiac function
  • Induces apoptosis in primary human lymphoblastic leukemia (ALL) cells
  • Delays growth of xenografts
  • Improves performance on behavioral tasks in Huntington disease models
Catalog No. 50-003-58394 Supplier Medchemexpress LLC Supplier No. HY50910200MG
Temsirolimus is an inhibitor of mTOR with an IC50 of 1.76 μM. It activates autophagy and prevents deterioration of cardiac function in animal models. This product is an analytical standard intended for research and analytical applications.
  • Inhibits mTOR kinase activity
  • Activates autophagy
  • Prevents deterioration of cardiac function
  • Induces apoptosis in primary human lymphoblastic leukemia (ALL) cells
  • Delays growth of xenografts
  • Improves performance on behavioral tasks in Huntington disease models