Learn More
Apexbio Technology LLC Temsirolimus 162635-04-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061706
Encompass_Preferred
Temsirolimus (CCI-779) is an ester analog of rapamycin functioning as an inhibitor of the mammalian target of rapamycin (mTOR) The compound suppresses mTOR signaling pathways thus inhibiting cellular proliferation and growth In preclinical studies on breast cancer cell lines Temsirolimus displayed inhibitory activities with reported IC50 values of 0 6 nM in BT-474 cells 0 7 nM in MDA-MB-468 and SKBR-3 cells and 50 nM in MCF-7 cells It has been examined in xenograft models of tumors with PTEN mutations and multiple myeloma for investigating pathways related to cell-cycle progression and proliferation regulated by mTOR
Catalog No. 50-000-61706 Supplier Apexbio Technology LLC Supplier No. A83145.1
 Restricted Item
Add to Cart
Add to Cart
Temsirolimus (CCI-779) is an ester analog of rapamycin functioning as an inhibitor of the mammalian target of rapamycin (mTOR) The compound suppresses mTOR signaling pathways thus inhibiting cellular proliferation and growth In preclinical studies on breast cancer cell lines Temsirolimus displayed inhibitory activities with reported IC50 values of 0 6 nM in BT-474 cells 0 7 nM in MDA-MB-468 and SKBR-3 cells and 50 nM in MCF-7 cells It has been examined in xenograft models of tumors with PTEN mutations and multiple myeloma for investigating pathways related to cell-cycle progression and proliferation regulated by mTOR