Learn More
Abcam Temsirolimus (CCI779), mTORC1 comple x inhibitor, 50MG
Catalog No. 502706379
Encompass_Preferred
MW 1030.3 Da, Purity >98%. Selective mTORC1 complex inhibitor (IC50 = 1.76 µM). Water-soluble rapamycin (ab120224) pro-drug. Rapidly converts to rapamycin *in vivo*. Inhibits proliferation and angiogenesis and induces apoptosis and autophagy *in vivo*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6379 Supplier Abcam Supplier No. AB14199950MG
MW 1030.3 Da, Purity >98%. Selective mTORC1 complex inhibitor (IC50 = 1.76 µM). Water-soluble rapamycin (ab120224) pro-drug. Rapidly converts to rapamycin *in vivo*. Inhibits proliferation and angiogenesis and induces apoptosis and autophagy *in vivo*.
The product is subject to the following: Abcam Restricted Use Statement