Learn More
Electron Microscopy Sciences TEM Window Silicon Dioxide, (8) 50 µm, (1) 50x100 µm, 20 nm
SDP
Catalog No. 501929473
Encompass_Preferred
By making the grids slightly narrower users now have easy access to grids in TEM holders. No more fumbling with tweezers while trying to pick up or put down grids. The new TEM Window grid shape is still compatible with all standard holders. TEM Window dimensions are 2.9mm in diameter and 100um thick.
Plasma Cleanable - can be vigorously plasma cleaned to remove organic contamination
Field to Field Uniformity - reduced variability
Tolerates temperatures above 1000°C - supports use in environmental TEMs where dynamic processes are observed at high temperatures
Withstands Harsh Conditions - Provides an ideal balance of imaging resolution, chemical stability and mechanical strength
Incorporates stoichiometric silicon dioxide - offers the ability to analyze for nitrogen by EDX techniques Lightly wrinkled with approximately 5um or less deflection in 100um of travel.
Dimensions: (8) 50 µm, (1) 50x100 µm
Thickness: 20 nm
Catalog No. 50-192-9473 Supplier Electron Microscopy Sciences Supplier No. 7604291
By making the grids slightly narrower users now have easy access to grids in TEM holders. No more fumbling with tweezers while trying to pick up or put down grids. The new TEM Window grid shape is still compatible with all standard holders. TEM Window dimensions are 2.9mm in diameter and 100um thick.
Plasma Cleanable - can be vigorously plasma cleaned to remove organic contamination
Field to Field Uniformity - reduced variability
Tolerates temperatures above 1000°C - supports use in environmental TEMs where dynamic processes are observed at high temperatures
Withstands Harsh Conditions - Provides an ideal balance of imaging resolution, chemical stability and mechanical strength
Incorporates stoichiometric silicon dioxide - offers the ability to analyze for nitrogen by EDX techniques Lightly wrinkled with approximately 5um or less deflection in 100um of travel.
Dimensions: (8) 50 µm, (1) 50x100 µm
Thickness: 20 nm